Anti-FAM117B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316163EA 543 EUR
ORIGTA316163
Anti-FAM117B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: FAM117B (family with sequence similarity 117, member B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR