Anti-HTR3D Rabbit Polyclonal Antibody

Supplier: OriGene
TA316222
ORIGTA316222EA 543 EUR
ORIGTA316222
Anti-HTR3D Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: HTR3D (5-hydroxytryptamine (serotonin) receptor 3D, ionotropic)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR