Anti-GPR115 Rabbit Polyclonal Antibody

Supplier: OriGene
TA316253
ORIGTA316253EA 564 EUR
ORIGTA316253
Anti-GPR115 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: GPR115 (G protein-coupled receptor 115)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR