Anti-NPAS4 Rabbit Polyclonal Antibody

Supplier: OriGene
TA316290
ORIGTA316290EA 564 EUR
ORIGTA316290
Anti-NPAS4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: NPAS4 (neuronal PAS domain protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR