Anti-OR10AG1 Rabbit Polyclonal Antibody

Supplier: OriGene
TA316295
ORIGTA316295EA 543 EUR
ORIGTA316295
Anti-OR10AG1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: OR10AG1 (olfactory receptor, family 10, subfamily AG, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR