Anti-SLC44A1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB5807-200EA 508 EUR
BIRBORB5807-200 BIRBORB5807-100
Anti-SLC44A1 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100; Immunochemistry: 1:100 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC44A1 (solute carrier family 44 (choline transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR