Imprimer
Anti-KIR2DS4 Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
BIRBORB5814-200EA 0 EUR
BIRBORB5814-200 BIRBORB5814-100
Anti-KIR2DS4 Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: KIR2DS4 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human