Anti-MO25 alpha Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB5906-200EA 508 EUR
BIRBORB5906-200 BIRBORB5906-100
Anti-MO25 alpha Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: MO25 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR