Imprimer

Anti-DHFR Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Biorbyt
BIRBORB5919-100EA 0 EUR
BIRBORB5919-100
Anti-DHFR Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: DHFR (dihydrofolate reductase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR