Anti-CUL4A Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB5921-20EA 109 EUR
BIRBORB5921-20 BIRBORB5921-200 BIRBORB5921-100 BIRBORB5921-5
Anti-CUL4A Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CUL4A (cullin 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR