Anti-COMP Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB5922-5EA 51 EUR
BIRBORB5922-5 BIRBORB5922-20 BIRBORB5922-200 BIRBORB5922-100
Anti-COMP Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: COMP (cartilage oligomeric matrix protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR