Anti-HBEGF Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB6014-100EA 312 EUR
BIRBORB6014-100 BIRBORB6014-200
Anti-HBEGF Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HBEGF (heparin-binding EGF-like growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR