Imprimer
Anti-HIST3H3 Rabbit polyclonal antibody
Fournisseur: Biorbyt
BIRBORB6154-100EA 0 EUR
BIRBORB6154-100
Anti-HIST3H3 Rabbit polyclonal antibody
Anticorps
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant