Anti-HIST1H3C Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB6167-100EA 306 EUR
BIRBORB6167-100 BIRBORB6167-20
Anti-HIST1H3C Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: HIST1H3C (histone cluster 1, H3c)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR