Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB21719-1EA 0 EUR
BIRBORB21719-1
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR