Anti-IgE Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21732-10EA 0 EUR
BIRBORB21732-10
Anti-IgE Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR