Anti-IgG Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21746-10EA 0 EUR
BIRBORB21746-10
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG (Duck) (duck IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR