Anti-IgA/IgG/IgM Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21852-1EA 0 EUR
BIRBORB21852-1
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgA/IgG/IgM (Chicken) (chicken IgA/IgG/IgM)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR