Anti-kappa light chain Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21881-1EA 0 EUR
BIRBORB21881-1
Anti-kappa light chain Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Mouse Ig Kappa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR