Imprimer

Anti-C3D Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB21912-1EA 0 EUR
BIRBORB21912-1
Anti-C3D Goat Polyclonal Antibody
Anticorps
Type: Secondary
Antigen: C3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR