Imprimer

Anti-LysozymeLysozyme Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: Biorbyt
BIRBORB21937-1EA 0 EUR
BIRBORB21937-1
Anti-LysozymeLysozyme Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Type: Secondary
Antigen: Lysozyme
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR