Anti-A1AT Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21955-1EA 0 EUR
BIRBORB21955-1
Anti-A1AT Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: alpha 1 Antitrypsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR