Anti- IgG3 lambda G3M (G) Antibody (Unconjugated)

Supplier: Biorbyt
BIRBORB22123-0.5EA 0 EUR
BIRBORB22123-0.5
Anti- IgG3 lambda G3M (G) Antibody (Unconjugated)
Antibodies
Type: Secondary
Antigen: IgG3 lambda G3M (G)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR