Anti- IgG4 kappa Gm4A Antibody (Unconjugated)

Supplier: Biorbyt
BIRBORB22126-0.5EA 0 EUR
BIRBORB22126-0.5
Anti- IgG4 kappa Gm4A Antibody (Unconjugated)
Antibodies
Type: Secondary
Antigen: IgG4 kappa Gm4A
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR