Anti-Mouse IgG1 IgG2A IgG2B IgG3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22160-1EA 0 EUR
BIRBORB22160-1
Anti-Mouse IgG1 IgG2A IgG2B IgG3 Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR