Anti-Monkey IgG Antibody (Unconjugated)

Supplier: Biorbyt
BIRBORB22199-10EA 0 EUR
BIRBORB22199-10
Anti-Monkey IgG Antibody (Unconjugated)
Antibodies
Type: Secondary
Antigen: Monkey(Rhesus) IgG
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR