Anti-Bovine Serum ProteinsBovine Serum Proteins Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22296-1EA 0 EUR
BIRBORB22296-1
Anti-Bovine Serum ProteinsBovine Serum Proteins Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Bovine Serum Proteins
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR