Anti-IgM Sheep Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22432-1EA 0 EUR
BIRBORB22432-1
Anti-IgM Sheep Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Guinea Pig IgM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR