Anti-Prealbumin Pig Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22464-1EA 0 EUR
BIRBORB22464-1
Anti-Prealbumin Pig Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Prealbumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Pig
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR