Anti-IgG Pig Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22467-1EA 0 EUR
BIRBORB22467-1
Anti-IgG Pig Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Pig
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR