Anti-SLC46A1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22475-100EA 0 EUR
BIRBORB22475-100
Anti-SLC46A1 Goat Polyclonal Antibody
Antibodies
ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: SLC46A1 (solute carrier family 46 (folate transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR