Anti-IRF2BP1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22523-100EA 0 EUR
BIRBORB22523-100
Anti-IRF2BP1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRF2BP1 (interferon regulatory factor 2 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR