Anti-CROT Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22526-100EA 0 EUR
BIRBORB22526-100
Anti-CROT Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CROT (carnitine O-octanoyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR