Anti-PRIM1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22527-100EA 0 EUR
BIRBORB22527-100
Anti-PRIM1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRIM1 (primase, DNA, polypeptide 1 (49kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR