Anti-FOXI3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22533-100EA 0 EUR
BIRBORB22533-100
Anti-FOXI3 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: FOXI3 (forkhead box I3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR