Anti-UGT2B Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22538-100EA 0 EUR
BIRBORB22538-100
Anti-UGT2B Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UGT2B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR