Anti-PNAS4 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22547-100EA 0 EUR
BIRBORB22547-100
Anti-PNAS4 Goat Polyclonal Antibody
Antibodies
ELISA: 1:32000; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PNAS4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR