Anti-GSTA5 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22558-100EA 0 EUR
BIRBORB22558-100
Anti-GSTA5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GSTA5 (glutathione S-transferase alpha 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR