Anti-NRG1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22617-20EA 0 EUR
BIRBORB22617-20 BIRBORB22617-5 BIRBORB22617-100
Anti-NRG1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: NRG1 (neuregulin 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR