Anti-YWHAG Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22801-100EA 0 EUR
BIRBORB22801-100 BIRBORB22801-20 BIRBORB22801-5
Anti-YWHAG Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: YWHAG (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR