Anti-HOMER1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22833-20EA 0 EUR
BIRBORB22833-20 BIRBORB22833-100 BIRBORB22833-5
Anti-HOMER1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HOMER1 (homer homolog 1 (Drosophila))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR