Anti-PDPN Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22898-5EA 0 EUR
BIRBORB22898-5 BIRBORB22898-100 BIRBORB22898-20
Anti-PDPN Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PDPN (podoplanin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR