Anti-SERPINB5 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22907-20EA 0 EUR
BIRBORB22907-20 BIRBORB22907-100 BIRBORB22907-5
Anti-SERPINB5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SERPINB5 (serpin peptidase inhibitor, clade B (ovalbumin), member 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR