Anti-Influenza A H1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB23032-100EA 0 EUR
BIRBORB23032-100
Anti-Influenza A H1 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1 ?g/ml

Type: Primary
Antigen: Influenza A H1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR