Anti-E. coli all ag's Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB23070-1EA 0 EUR
BIRBORB23070-1
Anti-E. coli all ag's Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: E. coli all ag's
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR