Anti-Neisseria gonorrhoeae Rabbit Polyclonal Antibody (Biotin)
Supplier: Biorbyt
BIRBORB23089-1EA 0 EUR
BIRBORB23089-1
Anti-Neisseria gonorrhoeae Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now