Anti-Treponema pallidum Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB23131-1EA 0 EUR
BIRBORB23131-1
Anti-Treponema pallidum Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Treponema pallidum
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR