Anti-Influenza A Goat Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB23142-1EA 0 EUR
BIRBORB23142-1
Anti-Influenza A Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Influenza A
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR