Anti-Hantavirus Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB23670-0.2EA 0 EUR
BIRBORB23670-0.2
Anti-Hantavirus Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Immunochemistry: 1:50; ELISA: 1:50; Western Blot: 1:50

Type: Primary
Antigen: Hantavirus
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR