Anti-Influenza A Nucleoprot. Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB23815-100EA 0 EUR
BIRBORB23815-100
Anti-Influenza A Nucleoprot. Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Immunofluorescence: 1:5-1:20

Type: Primary
Antigen: Influenza A Nucleoprot.
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR